Documentos clave
LIDF45645-EP
Lid for Eppendorf(R) FA-45-64-5-PCR Rotor
not aerosol-tight, for 5430/5430R, aluminum
About This Item
UNSPSC Code:
41103913
material
aluminum
feature
not aerosol-tight
manufacturer/tradename
Eppendorf® 022654225 (NA)
Eppendorf® 5427720006 (EU)
technique(s)
PCR: suitable
centrifugation: suitable
compatibility
for use with 5430 R
for use with 5430
Legal Information
Eppendorf is a registered trademark of Eppendorf AG