Key Documents
Z677825
Finntip® Flex pipette tips
Finntip Flex 10, non-sterile, pack of 1000 ea (Bag)
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
material
colorless tip
sterility
non-sterile
feature
barrier: no
packaging
pack of 1000 ea (Bag)
manufacturer/tradename
Finn 94060120
maximum volume
10 μL
volume range
0.1-10 μL
General description
Finntip Flex pipette tips are easy-to-use, soft, flexible high-end tips. They are molded with diamond polished tooling for an extra smooth tip surface, which prevents liquid retention and achieves better pipetting accuracy.
Other Notes
Browse all pipette tips
Legal Information
Finntip is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)