Key Documents
Z232289
Sigma-Aldrich
Filter/Washer/Degasser
capacity 100 mL, joint: ST/NS 14/20 M, F
About This Item
UNSPSC Code:
41100000
NACRES:
NB.43
label
CC
joint
joint: ST/NS14/20 M, F
capacity
100 mL
pore size
25-50 μm porosity
General description
Has septum-inlet, drip-tip, and PTFE stopcocks on side-arm, female top and male bottom joint.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Lot/Batch Number
Lo sentimos, en este momento no disponemos de COAs para este producto en línea.
Visite la Librería de documentos
Artículos
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.