Key Documents
Z101362
Sigma-Aldrich
Aldrich® septum-inlet adapter with stopcock
Plain, joint: ST/NS 19/22, PTFE stopcock
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
material
PTFE stopcock
joint
joint: ST/NS19/22
Categorías relacionadas
General description
Septum-Einlässe benötigen Septum Z10,072-2 oder Z12,435-4 sofern nicht mit * gekennzeichnet.
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)