Key Documents
CAD7910
Animal feeding needle, non-sterile, reusable, type 304 SS
curved, size 20G, L × diam. 1.5 in. × 2.25 mm , ball
About This Item
UNSPSC Code:
42142609
NACRES:
NB.03
size
20G
packaging
pkg of 12 ea
L × diam.
1.5 in. × 2.25 mm , ball
Features and Benefits
  • Fixed shape from stainless steel tubing
  • Stainless steel balls
  • Can be cleaned and reused as needed
  • Non-sterile
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)