Key Documents
Z742703
BenchMixer V2 Vortexer, foam rack for 12 x 15 mL tubes
About This Item
UNSPSC Code:
41103800
NACRES:
NB.73
AC/DC input
230 V
material
foamed PE
feature
Holds 12 x 15 mL tubes
packaging
pack of 1 ea
manufacturer/tradename
Benchmark Scientific BV1003-T150
General description
For use with BenchMixer V2 vortexer
Legal Information
Benchmixer is a trademark of Benchmark Scientific, Inc.