Key Documents
Z742638
Tube-O-DIALYZER Floats
Micro, MWCO 15 K
About This Item
UNSPSC Code:
41123100
NACRES:
NB.24
feature
Micro
packaging
pkg of 20 ea
manufacturer/tradename
G-BioSciences 786-613
pore size
15 K MWCO
storage temp.
2-8°C
General description
Replacement ”Tube-O-DIALYZER” floats
Legal Information
Tube-O-DIALYZER is a trademark of Geno Technology, Inc.