Documentos clave
Z601160
Hettich® MIKRO 220/220R centrifuge accessories
for MIKRO 220R Centrifuge, Adapter for 15 mL Falcon tubes (17x120 mm)
About This Item
UNSPSC Code:
41103911
NACRES:
NB.22
feature
Holds 15 mL tubes
packaging
pack of 6 ea
manufacturer/tradename
Hettich 1631
technique(s)
centrifugation: suitable
compatibility
for use with 1016, 1137 (rotors)
General description
  • compatible with rotor 1016 (Z601381)
  • rotor 1016 (Z601381) is only compatible with the MIKRO 220R Centrifuge
Legal Information
Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG
Required but not provided
Referencia del producto
Descripción
Precios
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)