Documentos clave
HS234521
Lab wipe holder
small, L × W × D 118 mm × 122 mm × 84 mm, clear acrylic
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
material
clear acrylic
feature
Holds Small wipes box
manufacturer/tradename
Heathrow Scientific HS234521
L × W × D
118 mm × 122 mm × 84 mm
General description
The clear acrylic Lab Wipe Holder holds Kimwipe and other major brand′s size boxes.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)