Documentos clave
GESE6119275
GE spacers
L × W × thickness 2 cm × 16 cm × 0.75 mm
About This Item
UNSPSC Code:
41105300
NACRES:
NB.22
packaging
pack of 2 ea
manufacturer/tradename
Cytiva Article No. 80-6180-51
Cytiva SE6119-2-,75
L × W × thickness
2 cm × 16 cm × 0.75 mm