Documentos clave
EP5820765003
Rotor for Eppendorf® 5804/5804R & 5810/5810R Centrifuges
FA-45-20-17, aerosol tight, to hold, 20 x 5.0 mL Eppendorf tubes, lid
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
feature
lid
to hold 20 x 5.0 mL Eppendorf tubes
aerosol tight
fixed angle rotor
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5820765003 (Global)
parameter
13,000 rpm
19083 × g max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5804/5804R/5810/5810R
General description
A wide selections of rotors to fit the Eppendorf 5804 and 5810 centrifuges.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)