Merck
EP0030122283
Eppendorf® Protein LoBind tubes
new
capacity 25 mL, PCR clean, screw top cap, speed 17,000 x g rpm, pkg of 200 ea (4 bags x 50 tubes)
About This Item
Código UNSPSC:
41121800
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Materiales
conical bottom
polypropylene
screw top cap
grado
for proteomics
descripción
withstand 17,000xg
esterilidad
non-sterile
Características
DNase free
PCR clean
RNase free
graduations: no
lid
envase
pkg of 200 ea (4 bags x 50 tubes)
fabricante / nombre comercial
Eppendorf® 0030122283
Parámetros
17,000 x g rpm speed
capacidad
25 mL
color
colorless
idoneidad
suitable for PCR
tipo de unión
Protein low binding
Descripción general
Avoid losing valuable samples during centrifugation, incubation and storage! Rely on the excellent protection offered by the hinged Safe-Lock lid. It contains all of our experience from 50 years of continuous optimization and development. Trust the original Eppendorf Safe-Lock Tubes, because your samples deserve only the best.
Protein LoBind Tubes, screw cap, Protein LoBind, 25 mL, PCR clean, colorless, 200 tubes (4 bags × 50 tubes)
Características y beneficios
  • Eppendorf LoBind material ensures optimized sample recovery for improved assay results
  • Free of surface coating (e.g., silicone) to minimize the risk of sample interference
  • Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors
  • Available in tube, microplate, and deepwell plate formats for easy-up scaling
  • Precise lid sealing to minimize evaporation
  • Rated up to 17,000_x_g centrifugation speed
Eppendorf Protein LoBind Tubes are specifically designed to ensure you get maximal protein recovery.
Información legal
Eppendorf is a registered trademark of Eppendorf AG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)