Documentos clave
Z532169
Sigma-Aldrich
Aldrich® single bank manifold with glass stopcocks
Manifold with vacuum-gauge port, positions, 3
About This Item
UNSPSC Code:
40161501
NACRES:
NB.79
feature
positions 3
L
300 mm
General description
Single bank manifolds come with 4 mm bore glass stopcocks.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)