Key Documents
Z171689
Ace double-jacketed coiled condenser
jacket L 250 mm
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
joint
joint ST/NS24/40
manufacturer/tradename
Ace Glass 595513
jacket L
250 mm
Categorías relacionadas
General description
Extra cooling capacity for volatile materials. Compact design retards diffusion.