Key Documents
SYNS311740
Sigma-Aldrich
Synthware T-bore glass stopcock Entire assembly
bore size 4 mm, plug size 17/40
Sinónimos:
Synthware S311740, t-shape stopcock
About This Item
UNSPSC Code:
41121800
NACRES:
NB.85
packaging
pkg of 1 ea
manufacturer/tradename
Synthware S311740
bore size
4 mm
Categorías relacionadas
General description
With solid glass plug. With constant pressure retention spring and clip.
Legal Information
Synthware is a trademark of Kemtech America Inc.