Key Documents
SYNC159190
Synthware West condenser
joint: ST/NS 19/22, jacket L 190 mm
Sinónimos:
Synthware C159190, glass West condenser
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
joint
joint: ST/NS19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C159190
jacket L
190 mm
Categorías relacionadas
General description
With narrow annular space for fast coolant speed, thus high cooling efficiency.
Legal Information
Synthware is a trademark of Kemtech America Inc.