Documentos clave
504351
Supelco
Conector Luer macho
configured for plugging luer holes, pkg of 12 ea
About This Item
Código UNSPSC:
12000000
envase
pkg of 12 ea
Nivel de calidad
técnicas
solid phase extraction (SPE): suitable
aplicaciones
environmental
food and beverages
compatibilidad
configured for plugging luer holes
Descripción general
Male Luer plug is used to seal the inlet sampling cap which serves as an accessory for thermal desorption system storage containers.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)