Documentos clave
Z332305
Bottle adapters for BRAND® Dispensers and Burettes
GL 45 thread, GL 38 bottle, polypropylene
About This Item
Código UNSPSC:
41121815
NACRES:
NB.01
Materiales
polypropylene
fabricante / nombre comercial
BRAND 704397
Adaptador
GL 38 bottle
GL 45 thread
Descripción general
For all Dispensette® models as well as seripettor® and QuikSip
Otras notas
BR704397 (India)
Información legal
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
QuikSip is a trademark of BRAND GMBH + CO KG
seripettor is a registered trademark of BRAND GMBH + CO KG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)