Z543586
Delivery cap for VerSA-Flow™ dispenser
three-hole
About This Item
Productos recomendados
material
PBT
PFA
PTFE
O.D.
1/8 in.
3.2 mm
fitting
GL 45 female
Categorías relacionadas
General description
Caps require a GL-45 bottle adapter such as Z332305 for use with Aldrich 2 L and 4 L bottles.
Legal Information
VerSA-Flow is a trademark of Sigma-Aldrich Co. LLC