Documentos clave
BMSC1005SC5S
Sigma-Aldrich
Microtube Centrifuge Tubes
capacity 5 mL, clear tube, sterile, cap (screw, attached), pack of 500 ea (10 x racks 50ea)
About This Item
UNSPSC Code:
41121703
NACRES:
NB.11
material
cap (screw, attached)
clear tube
conical bottom
polypropylene tube
sterility
sterile
feature
RNase and DNase free
graduations
packaging
pack of 500 ea (10 x racks 50ea)
manufacturer/tradename
Benchmark Scientific C1005-SC5-S
capacity
5 mL
tube diam.
16 mm
suitability
suitable for (centrifugation)
suitable for PCR
General description
5mL MacroTubes take all of the conveniences of standard 15mL conical tubes and scales them down for space efficiency and decreased natural resource consumption. Useful from -86°C to 110°C, these tubes are a valuable resource for boiling and for sample preservation at very low temperatures. Choose from gamma sterilized racked packaging with attached caps or non-sterile bulk packaging with separate caps. The foam racks are similar to the packaging normally associated with 15mL tubes (upright with caps attached), but they require about 60% less storage space.
Application
centrifugation, PCR