Documentos clave
XXSPTFF02
Millipore
Pellicon® Single-Pass TFF Cassette Diverter Plate
About This Item
Código UNSPSC:
23151800
eCl@ss:
32031690
Materiales
316L stainless steel housing
Nivel de calidad
técnicas
tangential flow filtration (TFF): suitable
aplicaciones
liquid formulation
Descripción general
The Pellicon Single-Pass TFF Cassette Diverter Plate is used to configure Pellicon 3 0.57 m², Pellicon 3 1.14 m² or Pellicon 2 0.5 m² cassettes for single-pass TFF operation.
Información legal
Pellicon is a registered trademark of Merck KGaA, Darmstadt, Germany