Documentos clave
CLS139545DC
Delivery cap for Corning® media storage bottles
red
Sinónimos:
Corning cap
About This Item
Código UNSPSC:
41121800
NACRES:
NB.43
fabricante / nombre comercial
Corning 1395-45DC
color
red
Categorías relacionadas
Descripción general
This three hole delivery cap has 3 holes with 1/4-28 threads at the top for use with 1/8in. O.D. tubing. No need to precut tubing to reach the reservoir bottom, just a quick quarter turn of the lock nut (from outside the reservoir) allows positioning of PTFE tubing at any depth. These delivery caps have a GL 45 thread, can be dry heat sterilized at up to 180 °C and have a PTFE liner for efficient sealing.
Información legal
Corning is a registered trademark of Corning, Inc.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)