Documentos clave
Z531901
Sigma-Aldrich
Tubing adapter with Torion connection and stopcock
PTFE stopcock, O.D. 1/2 in., male joint: ST/NS 24/40
Seleccione un Tamaño
1 EA
$239.000
$239.000
Fecha estimada de envío15 de abril de 2025
Seleccione un Tamaño
Cambiar Vistas
1 EA
$239.000
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
$239.000
Fecha estimada de envío15 de abril de 2025
material
PTFE stopcock
joint
male joint: ST/NS24/40
O.D.
1/2 in.
12.7 mm
General description
PTFE stopcocks are threaded for additional safety and have a 2 mm bore for 3/16 in. tubing connection and 4 mm bore for 5/16 in. and 1/2 in. tubing connections. 5/16in. O.D fits a #8 Torion, and 1/2in. O.D. fits a #14 Torion.
Legal Information
Torion is a trademark of Bibby Sterlin Ltd.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)