Key Documents
SYNV141003
Synthware conical bottom reaction vial
3 mL, joint: ST/NS 14/20
Sinónimos:
Synthware V141003, reaction tube
About This Item
UNSPSC Code:
41100000
NACRES:
NB.51
joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware V141003
capacity
3 mL
General description
Heavy wall vials have an internal conical bottom and flat bottom.
Legal Information
Synthware is a trademark of Kemtech America Inc.