Documentos clave
HAM238160
Hamilton® Single Pore lab pH electrode
glass, S7 connector
About This Item
UNSPSC Code:
41115604
NACRES:
NA.23
material
cylindrical H glass membrane
glass
single pore diaphragm
feature
autoclavable: no
electrode Skylyte-CL electrolyte
S7 connector
parameter
0-100 °C temp. range
diam.
12 mm
pH
0-14
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)