Documentos clave
ESYNTH006
Sigma-Aldrich
SynLectro 200 mL Jacketed Glass Cell with PTFE stopper
Seleccione un Tamaño
1 EACH
$2.079.000
$2.079.000
Fecha estimada de envío16 de abril de 2025
Solicitar un pedido a granel
Seleccione un Tamaño
Cambiar Vistas
1 EACH
$2.079.000
About This Item
UNSPSC Code:
81141502
NACRES:
NB.61
$2.079.000
Fecha estimada de envío16 de abril de 2025
Solicitar un pedido a granel
Categorías relacionadas
General description
SynLectro 200 mL Jacketed Glass Cell with PTFE Stopper is the part of the SynLectro Electrolysis Platform for synthetic electrochemistry.
Features and Benefits:
  • Designed to simplify setup, optimize reactivity, and improve safety
  • Side-arm enables connection to reflux condenser or inert gas
  • Jacket supports thermal control
  • Chemical resistant material (PTFE)
  • Protects electrical contacts from electrolytes
  • Sleeve (optional) prevents seized joints
SynLectro Electrolysis Platform
The SynLectro Electrolysis Platform is a portfolio of labware products and electrodes designed to simplify and standardize preparative scale electrosynthesis. Learn more about each component, designed with consistency, reproducibility, and safety in mind.
Legal Information
SynLectro is a trademark of Merck KGaA, Darmstadt, Germany
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Visite la Librería de documentos
Contenido relacionado
Learn about our new SynLectro™ Electrolysis Platform that has been designed to simplify and standardize electrochemical organic synthesis at research and batch scale.