Documentos clave
BR718905
BRAND® counting chamber BLAUBRAND® Bürker pattern
without clips, double ruled
Sinónimos:
BLAUBRAND®, Bürker, Cell culture, Counting Chambers
Seleccione un Tamaño
1 EA
$88.890
$88.890
Fecha estimada de envío29 de mayo de 2025
Seleccione un Tamaño
Cambiar Vistas
1 EA
$88.890
About This Item
Código UNSPSC:
41122600
NACRES:
NB.22
$88.890
Fecha estimada de envío29 de mayo de 2025
Materiales
colorless
glass
esterilidad
non-sterile
Características
without clips
fabricante / nombre comercial
BRAND 718905
pocillos
1
Descripción general
Manufactured according to DIN 12 847, including 2 haemacytometer cover glasses.The ruling shows 9 large squares of 1 mm² each. These are used for counting leucocytes.Each large square is subdivided by double lines (0.05 mm apart) into 16 group squares with 0.2 mm sides. The group squares correspond in size to the Neubauer counting chamber, but have no further subdivisions. They are used for counting thrombocytes and erythrocytes. The double lines from mini squares with an area of 0.0025 mm².
Características y beneficios
  • BLAUBRAND® quality
  • Outstanding measurement precision
  • 100% certified quality
Envase
1 counting chamber, complete with 2 haemacytometer cover glasses, in transparent plastic box.
Información legal
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)