Skip to Content
Merck

Skip To

C-243

C-Peptide solution

100 μg/mL (Protease-free 1x PBS), certified reference material, ampule of 0.5 mL, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C129H211N35O48
CAS Number:
Molecular Weight:
3020.26
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.24
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 0.5 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL (Protease-free 1x PBS)

format

single component solution

storage temp.

−20°C

SMILES string

N2([C@@H](CCC2)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCC(=O)O)C(=O)NCC(=O)N[C@@H](CO)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCC(=O)N)C(=O)O)C(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)CNC(=O)[C@@H](NC(=O)CNC(=O)[C@H]1N(CCC1)C(=O)CN

InChI

1S/C129H211N35O48/c1-58(2)42-77(155-113(195)74(31-39-102(184)185)151-127(209)105(65(15)16)162-114(196)71(25-32-88(131)167)145-95(174)54-141-126(208)104(64(13)14)161-115(197)72(26-33-89(132)168)150-118(200)80(45-61(7)8)156-121(203)83(48-103(186)187)159-112(194)73(30-38-101(182)183)148-107(189)67(18)143-109(191)69(130)24-36-99(178)179)111(193)138-50-93(172)135-49-92(171)136-55-98(177)163-40-20-22-86(163)124(206)140-51-94(173)142-66(17)106(188)137-52-96(175)146-84(56-165)122(204)157-81(46-62(9)10)119(201)152-75(27-34-90(133)169)128(210)164-41-21-23-87(164)125(207)160-78(43-59(3)4)116(198)144-68(19)108(190)154-79(44-60(5)6)117(199)149-70(29-37-100(180)181)110(192)139-53-97(176)147-85(57-166)123(205)158-82(47-63(11)12)120(202)153-76(129(211)212)28-35-91(134)170/h58-87,104-105,165-166H,20-57,130H2,1-19H3,(H2,131,167)(H2,132,168)(H2,133,169)(H2,134,170)(H,135,172)(H,136,171)(H,137,188)(H,138,193)(H,139,192)(H,140,206)(H,141,208)(H,142,173)(H,143,191)(H,144,198)(H,145,174)(H,146,175)(H,147,176)(H,148,189)(H,149,199)(H,150,200)(H,151,209)(H,152,201)(H,153,202)(H,154,190)(H,155,195)(H,156,203)(H,157,204)(H,158,205)(H,159,194)(H,160,207)(H,161,197)(H,162,196)(H,178,179)(H,180,181)(H,182,183)(H,184,185)(H,186,187)(H,211,212)/t66-,67-,68-,69-,70-,71-,72-,73-,74-,75-,76-,77-,78-,79-,80-,81-,82-,83-,84-,85-,86-,87-,104-,105-/m0/s1

InChI key

VOUAQYXWVJDEQY-QENPJCQMSA-N

General description

C-peptide, also known as the connecting peptide, is a 31-amino-acid polypeptide connecting insulin′s A-chain to its B-chain in the proinsulin molecule. Measurement of C-peptide blood serum levels can be used to distinguish between diabetes or hypoglycemia conditions with similar clinical features. This certified Snap-N-Spike® solution of the fully-intact c-peptide protein is suitable for use as starting material in calibrators and controls in LC-MS/MS c-peptide testing methods.
This certified reference material (CRM) is a primary standard. It is traceable to the SI and higher order standards from National Institute of Standards and Technology (NIST) through an unbroken chain of comparisons. This C-Peptide (human) acetate salt is available as a solution of a pure substance in phosphate buffered saline with a pH of 7.4.

Application

C-Peptide solution is suitable for analytical and R&D applications for in vitro identification, calibration, and quantification.

Features and Benefits

  • The total C-Peptide content is quantitatively determined using amino acid analysis (AAA) and HPLC/UV/MS, including unmodified, isomers, and pyroglu species, establishing traceability through the calibration of certification techniques.
  • Each raw material used in the preparation of this standard has undergone thorough identification and characterization using multiple analytical techniques.
  • The gravimetric preparation of this standard follows ISO 17025 requirements, utilizing qualified and calibrated balances.
  • Fill volume conformity is ensured throughout the dispensing process by gravimetric verification using qualified and calibrated balances.
  • Long-term stability under freezer storage (-10 °C to -25 °C) conditions has been evaluated.

Preparation Note

  • Thaw the product in the refrigerator or at room temperature.
  • Ensure thorough mixing before use.
  • Avoid refreezing the product once it has thawed.
  • For accurate results, quantitatively transfer the desired volume using established good laboratory practices.
  • Use the transferred product to spike into the matrix or dilute it to achieve the desired concentration.

Other Notes

This product′s purpose is for testing. THIS PRODUCT IS NOT INTENDED FOR HUMAN OR THERAPEUTIC USE. For comprehensive details regarding the nature of any hazards and recommended precautions, kindly refer to the safety data sheet.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
EZCCP-47KC4999EZHCP
manufacturer/tradename

Cerilliant®

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

grade

certified reference material

grade

-

grade

-

grade

-

format

single component solution

format

-

format

-

format

-

concentration

100 μg/mL (Protease-free 1x PBS)

concentration

-

concentration

-

concentration

-

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

Quality Level

300

Quality Level

200

Quality Level

200

Quality Level

200


Still not finding the right product?

Explore all of our products under C-Peptide solution


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
C-243-0.5ML04065266010275

Questions

Reviews

No rating value

Active Filters