Z188786
Wheaton sample vials
amber, volume 4 mL, cap size 13 - 425, diam. × H 15 mm × 46 mm
Synonym(s):
Wheaton Glass Vials
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
amber borosilicate glass vial
feature
closure type screw top vial
packaging
pkg of 200 ea
cap size
13 - 425
diam. × H
15 mm × 46 mm
volume
4 mL
color
amber
fitting
thread for 13-425
Looking for similar products? Visit Product Comparison Guide
General description
Vials are packaged in corrugated trays with partitions then shrink-wrapped to reduce particulate contamination.
Autoclavable borosilicate USP Type 1 glass. Order caps separately.
Autoclavable borosilicate USP Type 1 glass. Order caps separately.
related product
Product No.
Description
Pricing
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service