Z106526
Septa for open top caps
for cap size 13 mm, PTFE-faced styrene-butadiene rubber
All Photos(1)
About This Item
UNSPSC Code:
41122001
NACRES:
NB.11
Recommended Products
material
PTFE-faced styrene-butadiene rubber
packaging
pkg of 100
Related Categories
General description
For use with cut-out top caps and Wheaton vials.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)