Key Documents
Z106496
Supelco
Septa for open top caps
for cap size 13 mm, PTFE/silicone
About This Item
UNSPSC Code:
41122001
NACRES:
NB.11
material
PTFE/silicone
packaging
pkg of 100
General description
For use with cut-out top caps and Wheaton vials.
Choose from one of the most recent versions:
Certificates of Analysis (COA)