Documenti fondamentali
21950
Valco® Precision Sampling plunger tips
PTFE plunger tip, syringe size 500 μL, pkg of 2 ea
About This Item
Codice UNSPSC:
41122001
Materiali
PTFE
PTFE plunger tip
PTFE tip
Misura siringa
500 μL
Descrizione
plunger tip for 500μL syringe
Confezionamento
pkg of 2 ea
Produttore/marchio commerciale
VICI 013932
Misura
500 μL
Compatibilità
for use with Series A, A-2, C, & D Precision Sampling Syringe
Descrizione generale
For series A, A-2, C, and D Pressure-Lok Gas syringes. For 25 μL to 250 μL syringes, factory repair only.
Note legali
Valco is a registered trademark of Valco Instruments Co. Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Nandan S Gokhale et al.
Molecular cell, 77(3), 542-555 (2019-12-08)
The RNA modification N6-methyladenosine (m6A) modulates mRNA fate and thus affects many biological processes. We analyzed m6A across the transcriptome following infection by dengue virus (DENV), Zika virus (ZIKV), West Nile virus (WNV), and hepatitis C virus (HCV). We found