Documenti fondamentali
21804
Supelco
Empty Glass GC Column, for PerkinElmer® 8000, 9000, AutoSystem, XL, and Clarus 500 (Not On-Column)
L × O.D. × I.D. 6.0 ft (1.8 m) × 1/4 in. × 2.0 mm
About This Item
Codice UNSPSC:
41121800
Materiali
glass column
tecniche
gas chromatography (GC): suitable
Lungh. × D.E. × D.I.
6.0 ft (1.8 m) × 1/4 in. × 2.0 mm
Tipo di colonna
empty
Compatibilità
for use with PerkinElmer
Descrizione generale
GC columns are of two types; packed column and capillary column. Capillary column carries the gas through a central aperture which is unimpeded through the entire length of column. In general, capillary columns are made longer to produce more theoretical plates. This also helps in band dispersion eventually narrowing the peaks and resulting in high resolution.[1]
Silane treated empty glass column. Fittings are not included.
  • X (length of injector arm) = 6.81 in. (173 mm)
  • Y (length of detector arm) = 6.81 in. (173 mm)
  • S (span, injector to detector) = 6.5 in. (165 mm)
This column fits PerkinElmer 8000, 9000, AutoSystem, XL, and Clarus 500 (not on-column injection).
Applicazioni
Empty column may be used for packing adsorbents for temperature-programmed desorption (TPD) instrumentation.[2]
Note legali
PerkinElmer is a registered trademark of PerkinElmer Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
The determination of active filter aid adsorption sites by temperature-programmed desorption.
Zhu. Z. Y.
Journal of the American Oil Chemists' Society, 71, 189-194 (1994)
Raymond P. W. Scott
Introduction to Analytical Gas Chromatography (1997)