Z332232
Bottle adapters for BRAND® Dispensers and Burettes
GL 32 thread, S 40 bottle, polypropylene
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.01
Prodotti consigliati
Materiali
polypropylene
Produttore/marchio commerciale
BRAND 704343
Raccordi
S 40 bottle
GL 32 thread
Categorie correlate
Descrizione generale
For all Dispensette® models as well as seripettor® and QuikSip™
Altre note
BR704340 (India)
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
QuikSip is a trademark of BRAND GMBH + CO KG
seripettor is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)