Documenti fondamentali
TMO150318
Nunclon® cell culture dishes
diam. 35 mm, with lid, Nunclon® Delta treated, case of 500 ea
Sinonimo/i:
cell culture dish, cell culture treateddish
Scegli un formato
500 EACH
CHF 564.00
CHF 564.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
500 EACH
CHF 564.00
About This Item
Codice UNSPSC:
41121812
NACRES:
NB.13
CHF 564.00
Check Cart for Availability
Materiali
polystyrene dishes with lids
Sterilità
sterile; γ-irradiated
Confezionamento
case of 500 ea
Produttore/marchio commerciale
Nunc 150318
Diametro
35 mm
Descrizione generale
  • Large range available, treated for cell culture but with many other uses
  • Optically clear and very flat for use in microscope
  • Non-toxic
  • Certified surface treatment for optimal cell attachment and growth
Note legali
Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)