HS15994G
Premium Slide box
100 Place, cork lining, black
Sinonimo/i:
Schiebekasten, slide box microscope, slide storage, slide storage box
About This Item
Prodotti consigliati
Materiali
black
nickel closure
Caratteristiche
Holds 100 x slides 76x26 mm
Produttore/marchio commerciale
Heathrow Scientific HS15994G
Lungh. × Largh. × Alt.
208 × 175 × 34 mm
Categorie correlate
Descrizione generale
Caratteristiche e vantaggi
- molded, rigid boxes securely hold slides in grooved slots
- durable ABS plastic boxes are available wth either cork or foam lining
- both the lining and the slots are numbered to correspond to the slide inventory sheet
- closes securely with a rust-resistant, nickel-plated clasp and pin hinges
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)