Scegli un formato
| Voi/SKU | Disponibilità | Prezzo |
|---|
Informazioni su questo articolo
material
nickel closure, white plastic
feature
Holds 100 x slides 76x26 mm
manufacturer/tradename
Heathrow Scientific HS15994E
L × W × H
208 × 175 × 34 mm
Features and Benefits
- Molded, rigid boxes securely hold slides in grooved slots
- Durable ABS plastic boxes are available with cork lining only
- Both the lining and the slots are numbered to correspond to the slide inventory sheet
- Feature thumb-latch lock mechanism
1 of 1
Questo articolo | |||
|---|---|---|---|
| description 100 place, cork lining, white plastic | description 100 place, cork lining, gray plastic | description 100 place, foam, blue plastic | description 100 place, foam, red plastic |
| feature Holds 100 x slides 76x26 mm | feature Holds 100 x slides 76x26 mm | feature Holds 100 x slides 76x26 mm | feature Holds 100 x slides 76x26 mm |
| manufacturer/tradename Heathrow Scientific HS15994E | manufacturer/tradename Heathrow Scientific HS15994F | manufacturer/tradename Heathrow Scientific HS15994M | manufacturer/tradename Heathrow Scientific HS15994P |
| material white plastic , nickel closure | material nickel closure, gray plastic | material blue plastic , nickel closure | material nickel closure, red plastic |
| L × W × H 208 × 175 × 34 mm | L × W × H 208 × 175 × 34 mm | L × W × H 208 × 175 × 34 mm | L × W × H 208 × 175 × 34 mm |
Still not finding the right product?
Explore all of our products under Premium slide box
Scegli una delle versioni più recenti:
Possiedi già questo prodotto?