GN781097
Greiner microplate
384 well, flat μClear® bottom, high binding, black
Sinonimo/i:
384 well microplate, 384 well plate, cell culture plate
About This Item
Codice UNSPSC:
41121800
Prodotti consigliati
Materiali
black
Confezionamento
pack of 40 ea
Produttore/marchio commerciale
Greiner 781097
Disponibilità
available only in China
Misura
384 wells
Compatibilità
suitable for (cell culture)
Descrizione generale
- Greiner microplates are ideal for storing active agents, patient samples, DNA/RNA or stock cultures.
- Polypropylene material can withstand a high temperature (-196 to +121°C) and are chemically resistant.
- Nonpyrogenic plates are free of detectable DNase, RNase, human DNA.
- Features alphanumeric well coding for identification.
- Sealable with adhesive films and heat sealers for long-term and secure sealing.
Note legali
μClear is a registered trademark of Greiner Bio-One GmbH
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)