Documenti fondamentali
EP0030128680
Eppendorf® Twin tec PCR plates, 96 wells
red plate, conical bottom red wells, skirted, pkg of 25 ea
About This Item
Codice UNSPSC:
41106300
Materiali
conical bottom red wells
polycarbonate
red plate
Sterilità
non-sterile
Caratteristiche
skirt (Skirted)
Confezionamento
pkg of 25 ea
Produttore/marchio commerciale
Eppendorf® 0030128680
tecniche
PCR: suitable
Misura
96 wells
Volume massimo del pozzetto
150 μm
Descrizione generale
Choose Eppendorf twin.tec PCR plates for confidence in every PCR assay. twin.tec plates offer purity, rigidity, color, and other simple, smart features can make your daily life a lot easier. The rigid polycarbonate frame works well in automation and high-throughput processes, and the different colors mean you can quickly organize the different steps of your workflow.
Applicazioni
Eppendorf® Twin tec PCR plates, 96 wells are ideal for quantitative real-time PCR.
Caratteristiche e vantaggi
  • A combination of polypropylene wells and torque-resistant polycarbonate frame
  • DNA, DNAse, RNAse, and PCR inhibitors free
  • Extremely thin-walled polypropylene wells ensure optimal heat transfer to the samples
  • Alphanumeric grid-referencing and cut-away corner
  • Effective sealing and reduced risk of contamination due to raised well rims
  • Easy stacking
  • Upon request available with a barcode
  • Low-profile design enables high efficiency for small sample volumes and enhances PCR efficiency
  • Eight holes in the skirt aid plate and easy removal from the thermal cycler block
  • Compatible with automated systems
  • Support a snug fit to the PCR cycler and act as a solid base for tight sealing
Note legali
Eppendorf is a registered trademark of Eppendorf AG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)