Documenti fondamentali
CLS285018
Corning® cover glasses
square, No. 11/2, W × L 18 mm × 18 mm
Sinonimo/i:
coverslips, glass covers, slide covers
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.16
Prezzi e disponibilità al momento non sono disponibili
Materiali
borosilicate glass
square
Confezionamento
pack of 200 ea
case of 2000 ea
Produttore/marchio commerciale
Corning 2850-18
Largh. × lungh.
18 mm × 18 mm
Tipo di binding
non-treated surface
Caratteristiche e vantaggi
  • Corning® cover glasses are made from optically clear borosilicate glass.
  • Chemically resistant cover glasses are not affected by weathering or surface attachment.
Note legali
Corning is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 6
1 of 6
Corning® cover glasses square, No. 1, W x L 25 mm x 25 mm
CLS284525
Corning® cover glasses
Corning® cover glasses rectangular, No. 11/2, W × L 22 mm × 30 mm
CLS2980223
Corning® cover glasses
Corning® cover glasses rectangular, No. 11/2, W × L 22 mm × 40 mm
CLS2980224
Corning® cover glasses
BRAND® cover glass L × W 24 mm × 24 mm, square
BR470060
BRAND® cover glass
BRAND® cover glass L × W 22 mm × 22 mm, square
BR470055
BRAND® cover glass
Corning® cover glasses rectangular, No. 11/2, W × L 24 mm × 50 mm
CLS2980245
Corning® cover glasses