Z682373
Ace rotary evaporator condenser complete "V" assembly
glass, fits Büchi® models R-200/R-205 and R-114/R-144
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
Prodotti consigliati
Materiali
glass
Produttore/marchio commerciale
Büchi equivalent, 40601
Categorie correlate
Descrizione generale
The main glass condenser body with inner coil for vertical installation, O-ring, 1 L receiving flask, stainless steel clamp, GL threaded caps and hose barbs.
Note legali
Büchi is a registered trademark of Büchi Labortechnik AG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)