Documenti fondamentali
Z646202
Sigma-Aldrich
Height adjustable rack
Holds 6 x 1.5-15 mL tubes
Scegli un formato
1 EA
CHF 68.80
CHF 68.80
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 68.80
About This Item
Codice UNSPSC:
41122800
NACRES:
NB.35
CHF 68.80
Check Cart for Availability
Caratteristiche
Holds 6 x 1.5-15 mL tubes
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Bel-Art F18882-0000
Descrizione generale
Use the Scienceware Height-Adjustable Rack to securely hold up to six tubes or vials at any desired depth in a water bath or beaker. The rack has a flexible grip that easily retains mixed test tubes of varying weight and volume from 1.5 to 15 mL. Unlike floating racks, this rack′s adjustable height prevents submersion and sinking problems that might occur with heavier or unbalanced tubes.
The compact yet stable footprint saves bench and bath space and lifts in and out of a bath without splashing. All parts are steam autoclavable at 121ºC.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)