Documenti fondamentali
BAF187420001
PrepSafe Floating Microcentrifuge Tube Rack
Holds 28 x 1.5-2 mL tubes, polypropylene
Scegli un formato
1 EA
CHF 29.00
CHF 29.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 29.00
About This Item
Codice UNSPSC:
41122804
NACRES:
NB.22
CHF 29.00
Check Cart for Availability
Materiali
polypropylene
Caratteristiche
Holds 28 x 1.5-2 mL tubes
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Bel-Art F18742-0001
Misura
178x64x mm
Descrizione generale
Protect your samples. Hold 1.5ml and 2.0ml microcentrifuge tubes securely in a water bath or on the benchtop, even if tipped over. Immobilizing tabs allow tubes to be stored securely and ensure that the caps stay locked in place. This floating rack has 28 numbered locations so samples can easily be identified. Raised handle allows rack to be lifted without agitating samples. Patent pending.
Note legali
PrepSafe is a trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)