Documenti fondamentali
Z567744
PTFE-faced silicone septa for Q-Tube and QianCap
Septa 35 mL Low
Scegli un formato
100 EA
CHF 187.00
CHF 187.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
100 EA
CHF 187.00
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.51
CHF 187.00
Check Cart for Availability
Produttore/marchio commerciale
Q Labtech 100-0SEPTA-035
Parametri
120 psi max. pressure
diam. × spessore
30.2 mm × 2.5 mm
Descrizione generale
This disposable septa is made of blue PTFE face and white silicone backing. The blue PTFE liner has excellent chemical resistance and silicon backing has reseal ability. It fits on Q-Tube-12mL, QianCap-15 and 25. Using other septa may cause explosion.
Note legali
Q-Tube is a trademark of Q Labtech LLC
QianCap is a trademark of Q Labtech LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)