Documenti fondamentali
Z544566
Molyorbital set
Atomic Orbital
Scegli un formato
1 SET
CHF 44.90
CHF 44.90
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 SET
CHF 44.90
About This Item
Codice UNSPSC:
60104506
NACRES:
NB.17
CHF 44.90
Check Cart for Availability
Descrizione
14-model collection set
Produttore/marchio commerciale
Molymod
Descrizione generale
Contains 1s; 2s; 2p; 3d; 2s+ 3 × 2p; and sp, sp2, and sp3 hybridized models. Pink and purple lobes represent the +/- wave phases of the p and d atomic orbitals. Models come with transparent bases for display.
The Molymod molecular orbital system uses standard shiny colored atom parts to represent the central atom cores. Average scale is 3.5 cm per Angstrom. The atomic and molecular orbital parts are represented by pastel/matte colored pieces and are color-coded according to their use.
The pink and purple pear-shaped lobes and concave π links represent the +ve and -ve signs of the wave functions Ψ of the lobes of atomic and molecular orbitals. The gray pieces represent hybridized sigma bonds. The light brown spheres are used to indicate lone electron pair orbitals. Where the electrons are involved in hydrogen bonding, a light brown pear-shaped lobe is used
Note legali
Molyorbital is a trademark of Spiring Enterprises Ltd.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 3
1 of 3