Documenti fondamentali
Z544485
Molymod® molecular model set
teacher/researcher set, biochemistry
Scegli un formato
1 SET
CHF 117.00
CHF 117.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 SET
CHF 117.00
About This Item
Codice UNSPSC:
60104506
NACRES:
NB.17
CHF 117.00
Check Cart for Availability
Caratteristiche
no. of atoms 257
no. of bonds 160
Produttore/marchio commerciale
Molymod MMS-007
Descrizione generale
100 hydrogen atom-links, 10 hydrogen (ll), 68 carbon (ll, lll, lV), 40 oxygen (l, ll), 34 nitrogen (ll, lll, lV), 2 sulphur (ll), 6 phosphorus (lV), 2 metal (lV, Vl), Links: 150 non-vis colourless & 10 ‘V′-links
Top quality Molymod molecular model sets are for use by students, teachers, and researchers. May be upgraded or customized with additional model parts, for more info, go to sigma-aldrich.com and enter Molymod in our search engine.
Scale is 2.8 cm =100 pm (open style) and 1.45 cm = 100 pm (compact, semi-space-filling). Sets include standard color-coded plastic atom centers (with bond holes set at the appropriate angles), bonding links, and a plastic storage box.
Set pictured is representative of the Student set.
See our Selection Guide for detailed descriptions of set contents.
Note legali
Molymod is a registered trademark of Spiring Enterprises Ltd.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)