Documenti fondamentali
Z544558
Molyorbital set
Organic structures
Scegli un formato
1 SET
CHF 57.90
CHF 57.90
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 SET
CHF 57.90
About This Item
Codice UNSPSC:
60104506
NACRES:
NB.17
CHF 57.90
Check Cart for Availability
Descrizione
4-model collection set
Produttore/marchio commerciale
Molymod
Descrizione generale
Contains sufficient parts to make benzene, ethane, ethene, and ethyne. Shows sigma and pi bonding orbitals, and concepts of hybridization and delocalization.
The Molymod molecular orbital system uses standard shiny colored atom parts to represent the central atom cores. Average scale is 3.5 cm per Angstrom. The atomic and molecular orbital parts are represented by pastel/matte colored pieces and are color-coded according to their use.
The pink and purple pear-shaped lobes and concave π links represent the +ve and -ve signs of the wave functions Ψ of the lobes of atomic and molecular orbitals. The gray pieces represent hybridized sigma bonds. The light brown spheres are used to indicate lone electron pair orbitals. Where the electrons are involved in hydrogen bonding, a light brown pear-shaped lobe is used
Note legali
Molyorbital is a trademark of Spiring Enterprises Ltd.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)