Documenti fondamentali
Z268771
Safe-Lab Thermometer/Tubing Adapter
joint: ST/NS 10/18, opening size 6-7 mm
Sinonimo/i:
thermometer holder
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.35
Giunto
joint: ST/NS10/18
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Bel-Art F20309-1675
Dimens. apertura
6-7 mm
Descrizione generale
PTFE body with Viton® O-ring and PP screw-cap with inside Viton O-ring for air-tight work. Loosen cap for depth adjustment of thermometer or tubing.
Note legali
Safe-Lab is a trademark of SP Scienceware, a division of SP
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)