Documenti fondamentali
Z265942
Sigma-Aldrich
Solid-glass beads
soda-lime glass, diam. 5 mm
Scegli un formato
1 EA
CHF 34.10
CHF 34.10
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 34.10
About This Item
Codice UNSPSC:
41102422
NACRES:
NB.43
CHF 34.10
Check Cart for Availability
Materiali
soda-lime glass
Confezionamento
pack of 1 ea
Diametro
5 mm
Categorie correlate
Descrizione generale
For use as distillation column packing, mixing beads, and boiling stones. Each 454 g box has a packing volume of approx. 360 cm3. +/-10% variation in bead size.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 8
1 of 8
Glass beads 4 mm
Supelco
1.04016
Glass beads 4 mm
Soda-lime-silica glass NIST® SRM® 622
NIST622
Soda-lime-silica glass
Solid-glass beads borosilicate, diam. 3 mm
Z143928
Solid-glass beads
Glass beads diam. 0.5 mm, for Mini-Beadbeater
Sigma-Aldrich
Z250465
Glass beads
Glass beads 2 mm
Supelco
1.04014
Glass beads 2 mm
Glass beads 3 mm
Supelco
1.04015
Glass beads 3 mm
Glass beads, unwashed 425-600 μm (30-40 U.S. sieve)
G9268
Glass beads, unwashed