Documenti fondamentali
Z265934
Sigma-Aldrich
Solid-glass beads
soda-lime glass, diam. 4 mm
Scegli un formato
1 EA
CHF 36.70
CHF 36.70
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 36.70
About This Item
Codice UNSPSC:
41102422
NACRES:
NB.43
CHF 36.70
Check Cart for Availability
Materiali
soda-lime glass
Diametro
4 mm
Categorie correlate
Descrizione generale
For use as distillation column packing, mixing beads, and boiling stones. Each 454 g box has a packing volume of approx. 360 cm3. +/-10% variation in bead size.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 6
1 of 6
Questions
  1. How many beads are there in one pack? Sorry, I am unable to understand what 1 EA means.
    1 answer
    1. These beads have an average diameter of 4 mm. As listed in the DESCRIPTION section, the '1 EA' is a 454 g box that has a volume of approx. 360 mL +/- 10%. A bead count per package is not determined.